Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001173988.1 | NP_001167459.1 | 9 - 10 | 1401 | 127 |
NM_024718.4 | NP_078994.3 | 9 - 10 | 1398 | 127 | |
GenBank mRNA | AK000963.1 | - | 6 - 7 | 804 | 127 |
AK001184.1 | - | 8 - 9 | 985 | 127 | |
AK027586.1 | - | 6 - 7 | 819 | 127 | |
AK094382.1 | - | 9 - 10 | 1721 | 127 | |
AK098086.1 | - | 1 - 2 | 2489 | 127 | |
AK128154.2 | - | 9 - 10 | 1662 | 127 | |
AK307431.1 | - | 10 - 11 | 1252 | 127 | |
AL713707.1 | - | 2 - 3 | 374 | 127 | |
BC018960.2 | - | 9 - 10 | 1344 | 127 | |
BC021095.1 | - | 9 - 10 | 1102 | 127 | |
BC035786.1 | - | 9 - 10 | 1356 | 127 | |
DQ141240.1 | - | 9 - 10 | 1123 | 127 | |
EF156752.1 | - | 9 - 10 | 1125 | 127 |