Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199085.1 | NP_001186014.1 | 6 - 7 | 1487 | 125 |
NM_001199089.1 | NP_001186018.1 | 6 - 7 | 1200 | 125 | |
NM_001199091.1 | NP_001186020.1 | 6 - 7 | 1328 | 125 | |
NM_001199092.1 | NP_001186021.1 | 3 - 4 | 172 | 125 | |
NM_173533.3 | NP_775804.2 | 6 - 7 | 1487 | 125 | |
XM_005244934.1 | XP_005244991.1 | 6 - 7 | 1272 | 125 | |
XM_005244935.4 | XP_005244992.1 | 6 - 7 | 1177 | 125 | |
XM_017000473.1 | XP_016855962.1 | 6 - 7 | 1193 | 125 | |
GenBank mRNA | AK092142.1 | - | 3 - 4 | 187 | 125 |
AK126889.1 | - | 6 - 7 | 1327 | 125 | |
AK127370.1 | - | 6 - 7 | 1219 | 125 | |
BC065707.1 | - | 3 - 4 | 296 | 125 | |
BC089387.1 | - | 4 - 5 | 389 | 125 | |
BC130532.1 | - | 6 - 7 | 1164 | 125 | |
BC144058.1 | - | 6 - 7 | 1219 | 125 |