Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001253775.1 | NP_001240704.1 | 3 - 4 | 886 | 170 |
NM_001318246.1 | NP_001305175.1 | 3 - 4 | 473 | 170 | |
NM_194071.3 | NP_919047.2 | 3 - 4 | 886 | 170 | |
XM_005250541.4 | XP_005250598.1 | 3 - 4 | 1265 | 170 | |
XM_017012540.1 | XP_016868029.1 | 4 - 5 | 2288 | 170 | |
XM_017012541.1 | XP_016868030.1 | 4 - 5 | 851 | 170 | |
GenBank mRNA | AJ549092.1 | - | 3 - 4 | 839 | 170 |
AJ549387.1 | - | 3 - 4 | 839 | 170 | |
AK131517.1 | - | 3 - 4 | 852 | 170 | |
AK293048.1 | - | 3 - 4 | 818 | 170 | |
BC046181.1 | - | 2 - 3 | 347 | 170 | |
BC063666.1 | - | 3 - 4 | 829 | 170 | |
BC110813.1 | - | 3 - 4 | 822 | 170 | |
BX649143.1 | - | 3 - 4 | 478 | 170 |