Assay Design
Gene Transcripts
A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AF217973.1 |
- |
1 |
1143 |
|
Chr. 1 - 54104295 - 54106084 [+] on Build GRCh38 |
161 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
microRNA 4781 |
MIR4781 |
mir-4781 |
-50.1kb
|
Sense |
Chr. - 54054079 - 54054154 on Build GRCh38 |
Downstream |
CUB domain containing protein 2 |
CDCP2 |
|
32.9kb
|
Anti-sense |
Chr. - 54138995 - 54153006 on Build GRCh38 |
View Full Product Details