Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080456.2 | NP_001073925.2 | 1 - 2 | 384 | 96 |
XM_005258864.3 | XP_005258921.2 | 2 - 3 | 543 | 96 | |
XM_006723189.3 | XP_006723252.1 | 2 - 3 | 564 | 96 | |
GenBank mRNA | DW009806.1 | - | 1 - 2 | 252 | 96 |