Assay Design



Gene Transcripts

Gene Symbol
COL28A1
Entrez Gene ID
340267
Gene Name
collagen type XXVIII alpha 1 chain
Gene Aliases
COL28
Chromosome Location
Chr.7: 7338543 - 7542062 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037763.2 NP_001032852.2 3 - 4 795 145
XM_011515358.2 XP_011513660.1 3 - 4 5640 145
XM_011515359.2 XP_011513661.1 3 - 4 798 145
XM_011515360.2 XP_011513662.1 3 - 4 2080 145
XM_011515363.2 XP_011513665.1 3 - 4 2083 145
XM_011515364.2 XP_011513666.1 3 - 4 2082 145
XM_011515365.2 XP_011513667.1 3 - 4 2083 145
XM_011515366.2 XP_011513668.1 3 - 4 2083 145
XM_017012131.1 XP_016867620.1 4 - 5 6816 145
XM_017012132.1 XP_016867621.1 3 - 4 2080 145
XM_017012133.1 XP_016867622.1 3 - 4 2084 145
GenBank mRNA AJ890451.1 - 3 - 4 795 145
BC136892.1 - 3 - 4 785 145
BC144431.1 - 3 - 4 785 145

View Full Product Details