Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001010867.3 | NP_001010867.1 | 2 - 3 | 690 | 135 |
NM_001310327.1 | NP_001297256.1 | 2 - 3 | 401 | 135 | |
GenBank mRNA | AK022796.1 | - | 3 - 4 | 2717 | 135 |
AK303837.1 | - | 2 - 3 | 401 | 135 | |
BI820993.1 | - | 2 - 3 | 430 | 135 |