Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153214.2 | NP_694946.2 | 5 - 6 | 942 | 143 |
XM_006712260.3 | XP_006712323.1 | 5 - 6 | 942 | 143 | |
XM_011510585.2 | XP_011508887.1 | 5 - 6 | 942 | 143 | |
XM_011510587.2 | XP_011508889.1 | 6 - 7 | 919 | 143 | |
XM_017003317.1 | XP_016858806.1 | 5 - 6 | 942 | 143 | |
XM_017003318.1 | XP_016858807.1 | 5 - 6 | 942 | 143 | |
XM_017003319.1 | XP_016858808.1 | 5 - 6 | 942 | 143 | |
GenBank mRNA | AK094759.1 | - | 5 - 6 | 942 | 143 |
AK124067.1 | - | 3 - 4 | 339 | 143 | |
BC035784.1 | - | 5 - 6 | 895 | 143 |