Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017822.3 | NP_060292.3 | 3 - 4 | 509 | 139 |
GenBank mRNA | AK022732.1 | - | 2 - 3 | 247 | 139 |
AK094528.1 | - | 2 - 3 | 1032 | 139 | |
BC009746.2 | - | 3 - 4 | 477 | 139 | |
BC013900.2 | - | 2 - 3 | 429 | 139 |