Assay Design
Gene Transcripts
- Gene Symbol
- SRSF8
- Entrez Gene ID
-
10929
- Gene Name
- serine and arginine rich splicing factor 8
- Gene Aliases
-
DSM-1, SFRS2B, SRP46
- Chromosome Location
-
Chr.11: 95066877 - 95071227 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_103726.1 |
|
1 - 2 |
1308 |
107 |
View Full Product Details