Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080410.2 | NP_001073879.2 | 2 - 3 | 1130 | 152 |
XM_006711950.2 | XP_006712013.1 | 7 - 8 | 2114 | 152 | |
XM_011532562.1 | XP_011530864.1 | 3 - 4 | 1562 | 152 | |
XM_017003421.1 | XP_016858910.1 | 4 - 5 | 4211 | 152 | |
XM_017003422.1 | XP_016858911.1 | 7 - 8 | 3856 | 152 | |
XM_017003423.1 | XP_016858912.1 | 7 - 8 | 3237 | 152 | |
GenBank mRNA | AB075820.1 | - | 2 - 3 | 968 | 152 |