Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080410.2 | NP_001073879.2 | 7 - 8 | 2072 | 167 |
XM_005264158.3 | XP_005264215.1 | 5 - 6 | 991 | 167 | |
XM_006711950.2 | XP_006712013.1 | 12 - 13 | 3056 | 167 | |
XM_011532562.1 | XP_011530864.1 | 8 - 9 | 2504 | 167 | |
XM_011532563.2 | XP_011530865.1 | 6 - 7 | 1429 | 167 | |
XM_017003421.1 | XP_016858910.1 | 9 - 10 | 5153 | 167 | |
XM_017003422.1 | XP_016858911.1 | 12 - 13 | 4798 | 167 | |
XM_017003423.1 | XP_016858912.1 | 12 - 13 | 4179 | 167 | |
GenBank mRNA | AB075820.1 | - | 7 - 8 | 1910 | 167 |
AK127503.1 | - | 4 - 5 | 553 | 167 | |
BC112202.1 | - | 6 - 7 | 980 | 167 |