Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000497.3 | NP_000488.3 | 2 - 3 | 398 | 137 |
NM_001026213.1 | NP_001021384.1 | 2 - 3 | 398 | 137 | |
GenBank mRNA | AF478474.1 | - | 2 - 3 | 398 | 137 |
BC096286.3 | - | 2 - 3 | 398 | 137 | |
BC096287.3 | - | 2 - 3 | 398 | 137 | |
X55764.1 | - | 2 - 3 | 395 | 137 |