Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278689.1 | NP_001265618.1 | 14 - 15 | 1830 | 141 |
NM_173654.2 | NP_775925.1 | 11 - 12 | 1578 | 141 | |
NR_103826.1 | 12 - 13 | 1619 | 141 | ||
XM_005264743.2 | XP_005264800.1 | 14 - 15 | 2044 | 141 | |
XM_011533599.1 | XP_011531901.1 | 15 - 16 | 2098 | 141 | |
XM_011533600.1 | XP_011531902.1 | 15 - 16 | 1951 | 141 | |
XM_011533602.1 | XP_011531904.1 | 15 - 16 | 1863 | 141 | |
XM_011533603.1 | XP_011531905.1 | 15 - 16 | 2095 | 141 | |
XM_017006204.1 | XP_016861693.1 | 15 - 16 | 1948 | 141 | |
XM_017006205.1 | XP_016861694.1 | 14 - 15 | 1809 | 141 | |
XM_017006206.1 | XP_016861695.1 | 14 - 15 | 1705 | 141 | |
XM_017006207.1 | XP_016861696.1 | 7 - 8 | 544 | 141 | |
XM_017006209.1 | XP_016861698.1 | 5 - 6 | 417 | 141 | |
GenBank mRNA | AJ868234.1 | - | 11 - 12 | 1154 | 141 |
AK091089.1 | - | 5 - 6 | 808 | 141 | |
AK123213.1 | - | 2 - 3 | 142 | 141 | |
AK290356.1 | - | 10 - 11 | 957 | 141 | |
AK294101.1 | - | 6 - 7 | 470 | 141 | |
AK304102.1 | - | 12 - 13 | 1375 | 141 | |
BC060887.1 | - | 11 - 12 | 1307 | 141 | |
BX640821.1 | - | 10 - 11 | 951 | 141 | |
BX648818.1 | - | 14 - 15 | 1605 | 141 | |
KC347596.1 | - | 11 - 12 | 1154 | 141 |