Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001077619.1 | NP_001071087.1 | 3 - 4 | 462 | 78 |
XM_005251165.2 | XP_005251222.1 | 3 - 4 | 365 | 78 | |
XM_005251166.1 | XP_005251223.1 | 3 - 4 | 365 | 78 | |
GenBank mRNA | AK126300.1 | - | 3 - 4 | 365 | 78 |
AL831990.2 | - | 3 - 4 | 336 | 78 | |
AL833075.2 | - | 3 - 4 | 336 | 78 | |
BC113645.1 | - | 3 - 4 | 462 | 78 | |
BX647876.2 | - | 3 - 4 | 336 | 78 |