Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001004312.2 | NP_001004312.2 | 1 - 2 | 593 | 106 |
XM_017006301.1 | XP_016861790.1 | 4 - 5 | 787 | 106 | |
XM_017006302.1 | XP_016861791.1 | 4 - 5 | 738 | 106 | |
GenBank mRNA | AY562236.1 | - | 1 - 2 | 164 | 106 |
BC068081.1 | - | 1 - 2 | 593 | 106 |