Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002777.3 | NP_002768.3 | 4 - 5 | 628 | 102 |
GenBank mRNA | BC096183.1 | - | 4 - 5 | 639 | 102 |
BC096184.3 | - | 4 - 5 | 639 | 102 | |
BC096185.3 | - | 4 - 5 | 639 | 102 | |
BC096186.3 | - | 4 - 5 | 639 | 102 | |
FN647680.1 | - | 3 - 4 | 663 | 102 | |
M29142.1 | - | 3 - 4 | 547 | 102 | |
M75154.1 | - | 4 - 5 | 652 | 102 | |
X55668.1 | - | 4 - 5 | 601 | 102 | |
X56132.1 | - | 3 - 4 | 549 | 102 |