Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004075.4 | NP_004066.1 | 8 - 9 | 2191 | 104 |
XM_017018832.1 | XP_016874321.1 | 9 - 10 | 2326 | 104 | |
GenBank mRNA | AK125915.1 | - | 10 - 11 | 1854 | 104 |
AK290552.1 | - | 8 - 9 | 1491 | 104 | |
BC017466.1 | - | 8 - 9 | 1614 | 104 | |
BC030519.1 | - | 8 - 9 | 1864 | 104 | |
D83702.1 | - | 8 - 9 | 1881 | 104 | |
D84657.1 | - | 8 - 9 | 1295 | 104 |