Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003981.3 | NP_003972.1 | 2 - 3 | 377 | 149 |
NM_199413.2 | NP_955445.1 | 2 - 3 | 377 | 149 | |
XM_005254987.2 | XP_005255044.1 | 2 - 3 | 285 | 149 | |
XM_006720759.1 | XP_006720822.1 | 2 - 3 | 285 | 149 | |
XM_006720760.1 | XP_006720823.1 | 2 - 3 | 285 | 149 | |
XM_011522187.1 | XP_011520489.1 | 2 - 3 | 285 | 149 | |
XM_011522188.2 | XP_011520490.1 | 2 - 3 | 285 | 149 | |
XM_011522189.1 | XP_011520491.1 | 2 - 3 | 285 | 149 | |
XM_011522190.2 | XP_011520492.1 | 2 - 3 | 285 | 149 | |
XM_011522191.2 | XP_011520493.1 | 2 - 3 | 285 | 149 | |
XM_011522192.1 | XP_011520494.1 | 2 - 3 | 285 | 149 | |
XM_017022712.1 | XP_016878201.1 | 2 - 3 | 285 | 149 | |
XM_017022713.1 | XP_016878202.1 | 2 - 3 | 285 | 149 | |
XM_017022716.1 | XP_016878205.1 | 2 - 3 | 285 | 149 | |
GenBank mRNA | AF044588.1 | - | 2 - 3 | 221 | 149 |
AK312600.1 | - | 2 - 3 | 276 | 149 | |
BC003138.1 | - | 2 - 3 | 245 | 149 | |
BC005140.2 | - | 2 - 3 | 275 | 149 | |
BG773441.1 | - | 2 - 3 | 333 | 149 | |
BX457401.2 | - | 2 - 3 | 230 | 149 | |
DA655494.1 | - | 2 - 3 | 378 | 150 |