Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_194449.3 | NP_919431.2 | 15 - 16 | 3997 | 160 |
GenBank mRNA | AB011178.2 | - | 15 - 16 | 2710 | 160 |
BC010706.1 | - | 2 - 3 | 232 | 160 | |
BC014927.2 | - | 15 - 16 | 2284 | 160 | |
BC047653.1 | - | 4 - 5 | 571 | 160 | |
BC063519.1 | - | 3 - 4 | 445 | 160 | |
BC126277.1 | - | 15 - 16 | 2332 | 160 |