Assay Design



Gene Transcripts

Gene Symbol
SUCLA2
Entrez Gene ID
8803
Gene Name
succinate-CoA ligase ADP-forming beta subunit
Gene Aliases
A-BETA, MTDPS5, SCS-betaA
Chromosome Location
Chr.13: 47942656 - 48037972 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_017020807.1 XP_016876296.1 2 - 3 293 134
XM_017020809.1 XP_016876298.1 2 - 3 318 134
GenBank mRNA AK302311.1 - 2 - 3 293 134

View Full Product Details