Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003850.2 | NP_003841.1 | 4 - 5 | 588 | 93 |
XM_017020806.1 | XP_016876295.1 | 4 - 5 | 808 | 93 | |
XM_017020807.1 | XP_016876296.1 | 6 - 7 | 823 | 93 | |
XM_017020808.1 | XP_016876297.1 | 4 - 5 | 575 | 93 | |
XM_017020809.1 | XP_016876298.1 | 5 - 6 | 672 | 93 | |
GenBank mRNA | AB035863.1 | - | 4 - 5 | 588 | 93 |
AF058953.1 | - | 3 - 4 | 420 | 93 | |
AF087890.1 | - | 4 - 5 | 596 | 93 | |
AK001458.1 | - | 4 - 5 | 486 | 93 | |
AK001847.1 | - | 4 - 5 | 575 | 93 | |
AK225452.1 | - | 4 - 5 | 575 | 93 | |
AK302311.1 | - | 5 - 6 | 647 | 93 | |
AK315513.1 | - | 4 - 5 | 550 | 93 | |
AK316271.1 | - | 5 - 6 | 903 | 93 | |
BC027587.2 | - | 4 - 5 | 540 | 93 | |
BX640602.1 | - | 4 - 5 | 559 | 93 | |
KU178442.1 | - | 4 - 5 | 531 | 93 |