Assay Design
Gene Transcripts
- Gene Symbol
- MED27
- Entrez Gene ID
-
9442
- Gene Name
- mediator complex subunit 27
- Gene Aliases
-
CRAP34, CRSP34, CRSP8, MED3, TRAP37
- Chromosome Location
-
Chr.9: 131860110 - 132079887 on Build GRCh38
- Gene Symbol
- CRSP8P
- Entrez Gene ID
-
441089
- Gene Name
- mediator complex subunit 27 pseudogene
- Gene Aliases
-
-
- Chromosome Location
-
Chr.5: 80350605 - 80351966 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC119722.1 |
- |
3 - 3 |
431 |
83 |
BC119723.1 |
- |
3 - 3 |
431 |
83 |
View Full Product Details