Assay Design
Gene Transcripts
- Gene Symbol
- GPR18
- Entrez Gene ID
-
2841
- Gene Name
- G protein-coupled receptor 18
- Gene Aliases
-
-
- Chromosome Location
-
Chr.13: 99254713 - 99262498 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_005292.3 |
NP_005283.1 |
1 - 2 |
271 |
93 |
GenBank mRNA |
BC050646.1 |
- |
1 - 2 |
217 |
93 |
View Full Product Details