Assay Design



Gene Transcripts

Gene Symbol
GLUD2
Entrez Gene ID
2747
Gene Name
glutamate dehydrogenase 2
Gene Aliases
GDH2, GLUDP1
Chromosome Location
Chr.X: 121047608 - 121049942 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012084.3 NP_036216.2 1 - 1 144 92
GenBank mRNA AK313356.1 - 1 - 1 141 92
BC050732.2 - 1 - 1 144 92

View Full Product Details