Assay Design



Gene Transcripts

Gene Symbol
RAB6C
Entrez Gene ID
84084
Gene Name
RAB6C, member RAS oncogene family
Gene Aliases
WTH3
Chromosome Location
Chr.2: 129979662 - 129982738 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032144.2 NP_115520.2 1 - 1 2239 116
GenBank mRNA AK055504.1 - 2 - 2 1619 109
AL136727.1 - 1 - 1 2233 108

View Full Product Details