Assay Design



Gene Transcripts

Gene Symbol
VHL
Entrez Gene ID
7428
Gene Name
von Hippel-Lindau tumor suppressor
Gene Aliases
HRCA1, RCA1, VHL1, pVHL
Chromosome Location
Chr.3: 10141635 - 10153670 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198156.2 NP_937799.1 1 - 2 551 112
GenBank mRNA BX100230.1 - 1 - 2 112
KU178386.1 - 1 - 2 338 112

View Full Product Details