Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001306084.1 | NP_001293013.1 | 54 - 55 | 7597 | 169 |
XM_011539069.2 | XP_011537371.1 | 55 - 56 | 7797 | 169 | |
XM_011539070.2 | XP_011537372.1 | 55 - 56 | 7797 | 169 | |
XM_011539071.2 | XP_011537373.1 | 55 - 56 | 7797 | 169 | |
XM_011539072.2 | XP_011537374.1 | 55 - 56 | 7797 | 169 | |
XM_011539073.2 | XP_011537375.1 | 55 - 56 | 7797 | 169 | |
XM_011539074.2 | XP_011537376.1 | 55 - 56 | 7797 | 169 | |
XM_011539076.2 | XP_011537378.1 | 55 - 56 | 7797 | 169 | |
XM_011539077.2 | XP_011537379.1 | 54 - 55 | 7947 | 169 | |
XM_011539078.2 | XP_011537380.1 | 55 - 56 | 7797 | 169 | |
XM_011539079.1 | XP_011537381.1 | 51 - 52 | 7043 | 169 | |
XM_011539081.2 | XP_011537383.1 | 55 - 56 | 7797 | 169 | |
XM_011539083.2 | XP_011537385.1 | 55 - 56 | 7797 | 169 | |
XM_011539085.1 | XP_011537387.1 | 40 - 41 | 5618 | 169 | |
XM_017018866.1 | XP_016874355.1 | 55 - 56 | 7797 | 169 | |
XM_017018867.1 | XP_016874356.1 | 44 - 45 | 6049 | 169 | |
GenBank mRNA | AK126100.1 | - | 21 - 22 | 2911 | 169 |