Assay Design
Gene Transcripts
- Gene Symbol
- HFM1
- Entrez Gene ID
-
164045
- Gene Name
- HFM1, ATP dependent DNA helicase homolog
- Gene Aliases
-
MER3, POF9, SEC63D1, Si-11, Si-11-6, helicase
- Chromosome Location
-
Chr.1: 91260766 - 91408008 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK094079.1 |
- |
1 - 2 |
132 |
108 |
View Full Product Details