Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291661.1 | NP_001278590.1 | 8 - 9 | 1078 | 150 |
NM_198564.3 | NP_940966.2 | 8 - 9 | 1078 | 150 | |
XM_011533466.1 | XP_011531768.1 | 8 - 9 | 1075 | 150 | |
XM_011533467.1 | XP_011531769.1 | 8 - 9 | 1042 | 150 | |
XM_011533468.1 | XP_011531770.1 | 8 - 9 | 1075 | 150 | |
XM_011533469.1 | XP_011531771.1 | 8 - 9 | 1075 | 150 | |
XM_011533471.2 | XP_011531773.1 | 8 - 9 | 1075 | 150 | |
XM_011533474.1 | XP_011531776.1 | 8 - 9 | 1075 | 150 | |
XM_017005860.1 | XP_016861349.1 | 8 - 9 | 1075 | 150 | |
XM_017005861.1 | XP_016861350.1 | 8 - 9 | 1937 | 150 | |
XM_017005863.1 | XP_016861352.1 | 8 - 9 | 1075 | 150 | |
GenBank mRNA | AK126276.1 | - | 8 - 9 | 1075 | 150 |