Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291661.1 | NP_001278590.1 | 9 - 10 | 1266 | 79 |
NM_198564.3 | NP_940966.2 | 9 - 10 | 1266 | 79 | |
XM_011533466.1 | XP_011531768.1 | 9 - 10 | 1263 | 79 | |
XM_011533467.1 | XP_011531769.1 | 9 - 10 | 1230 | 79 | |
XM_011533468.1 | XP_011531770.1 | 9 - 10 | 1263 | 79 | |
XM_011533469.1 | XP_011531771.1 | 9 - 10 | 1263 | 79 | |
XM_011533471.2 | XP_011531773.1 | 9 - 10 | 1263 | 79 | |
XM_011533474.1 | XP_011531776.1 | 9 - 10 | 1263 | 79 | |
XM_017005860.1 | XP_016861349.1 | 9 - 10 | 1263 | 79 | |
XM_017005861.1 | XP_016861350.1 | 9 - 10 | 2125 | 79 | |
XM_017005863.1 | XP_016861352.1 | 9 - 10 | 1263 | 79 | |
GenBank mRNA | AK126276.1 | - | 9 - 10 | 1263 | 79 |