Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_034156.1 | 3 - 4 | 347 | 98 | |
GenBank mRNA | AK123499.1 | - | 3 - 4 | 347 | 98 |
BC128606.1 | - | 2 - 3 | 251 | 98 | |
FJ613353.1 | - | 3 - 4 | 301 | 98 | |
FJ613354.1 | - | 3 - 4 | 328 | 98 |