Assay Design



Gene Transcripts

Gene Symbol
TPTE2
Entrez Gene ID
93492
Gene Name
transmembrane phosphoinositide 3-phosphatase and tensin homolog 2
Gene Aliases
TPIP
Chromosome Location
Chr.13: 19422877 - 19561574 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_073486.1 3 - 4 163 152
GenBank mRNA AJ421033.1 - 2 - 3 120 152

View Full Product Details