Assay Design



Gene Transcripts

Gene Symbol
TPTE2
Entrez Gene ID
93492
Gene Name
transmembrane phosphoinositide 3-phosphatase and tensin homolog 2
Gene Aliases
TPIP
Chromosome Location
Chr.13: 19422877 - 19561574 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001141968.1 NP_001135440.1 11 - 12 950 169
NM_130785.3 NP_570141.3 10 - 11 860 169
NM_199254.2 NP_954863.2 13 - 14 1091 169
NR_073485.1 15 - 16 1203 169
NR_073486.1 8 - 9 530 169
GenBank mRNA AJ421032.1 - 10 - 11 860 169
AJ421033.1 - 7 - 8 487 169
AK301741.1 - 15 - 16 1203 169
AY219890.1 - 13 - 14 1091 169
BC128146.1 - 11 - 12 932 169

View Full Product Details