Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001244767.1 | NP_001231696.1 | 1 - 2 | 212 | 74 |
NM_173468.3 | NP_775739.1 | 1 - 2 | 212 | 74 | |
GenBank mRNA | AK093545.1 | - | 1 - 2 | 205 | 74 |
AK313513.1 | - | 1 - 2 | 97 | 74 | |
BC038112.1 | - | 1 - 2 | 165 | 74 |