Assay Design



Gene Transcripts

Gene Symbol
ERICH5
Entrez Gene ID
203111
Gene Name
glutamate rich 5
Gene Aliases
C8orf47
Chromosome Location
Chr.8: 98064522 - 98093610 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001170806.1 NP_001164277.1 1 - 2 209 79
GenBank mRNA AI697720.1 - 1 - 2 79

View Full Product Details