Assay Design



Gene Transcripts

Gene Symbol
OR52D1
Entrez Gene ID
390066
Gene Name
olfactory receptor family 52 subfamily D member 1
Gene Aliases
OR11-43
Chromosome Location
Chr.11: 5488685 - 5489749 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005163.2 NP_001005163.1 1 - 1 931 82
GenBank mRNA BC136928.1 - 1 - 1 931 82
BC136930.1 - 1 - 1 931 82

View Full Product Details