Assay Design
Gene Transcripts
- Gene Symbol
- SEPT7P2
- Entrez Gene ID
-
641977
- Gene Name
- septin 7 pseudogene 2
- Gene Aliases
-
SEPT13, SEPT7B
- Chromosome Location
-
Chr.7: 45723787 - 45769018 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AL832059.1 |
- |
10 - 11 |
1214 |
122 |
View Full Product Details