Assay Design



Gene Transcripts

Gene Symbol
PRAMEF20
Entrez Gene ID
645425
Gene Name
PRAME family member 20
Gene Aliases
PRAMEF21
Chromosome Location
Chr.1: 13410450 - 13421328 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099852.2 NP_001093322.2 3 - 4 960 158

View Full Product Details