Assay Design



Gene Transcripts

Gene Symbol
PRAMEF17
Entrez Gene ID
391004
Gene Name
PRAME family member 17
Gene Aliases
-
Chromosome Location
Chr.1: 13389632 - 13392629 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099851.3 NP_001093321.1 2 - 3 895 163
GenBank mRNA BC144662.1 - 2 - 3 895 163
BC146858.1 - 2 - 3 895 163

View Full Product Details