Assay Design
Gene Transcripts
- Gene Symbol
- SKAP2
- Entrez Gene ID
-
8935
- Gene Name
- src kinase associated phosphoprotein 2
- Gene Aliases
-
PRAP, RA70, SAPS, SCAP2, SKAP-HOM, SKAP55R
- Chromosome Location
-
Chr.7: 26654772 - 26864743 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AF174049.1 |
- |
1 - 1 |
151 |
178 |
View Full Product Details