Product Details

Assay Reference Genome
Location

Chr.9:35075554 on build GRCh38
Cytoband
9p13.3
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
2189
Gene Symbol
FANCG
Gene Name
Fanconi anemia complementation group G
Gene Aliases
FAG, XRCC9
Location
Chr.9:35073838-35080016 on build GRCh38
Assay Gene Location
Within Exon 10
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
FANCG NM_004629.1 10 1836 NP_004620.1
AJ007669.1 10 1836 CAA07602.1
AK293427.1
AK311348.1 9 1310
AK312987.1 10 1761
BC000032.2 10 1482 AAH00032.1
BC011623.2 10 1609 AAH11623.1
BT009813.1 10 1344 AAP88815.1
KU159422.1 10 1849
KU178053.1 10 1344
KU178054.1 10 1326
KU178055.1 10 1344
KU178056.1 7 918
KU178057.1
U70310.1 10 1478 AAB80802.1

View Full Product Details