Assay Design
Gene Transcripts
- Gene Symbol
- LINC00471
- Entrez Gene ID
-
151477
- Gene Name
- long intergenic non-protein coding RNA 471
- Gene Aliases
-
C2orf52
- Chromosome Location
-
Chr.2: 231508426 - 231514339 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_024079.1 |
|
3 - 3 |
728 |
119 |
GenBank mRNA |
BC033054.2 |
- |
3 - 3 |
728 |
119 |
View Full Product Details