Assay Design
Gene Transcripts
- Gene Symbol
- MT1B
- Entrez Gene ID
-
4490
- Gene Name
- metallothionein 1B
- Gene Aliases
-
MT1, MT1Q, MTP
- Chromosome Location
-
Chr.16: 56651899 - 56653204 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_005947.2 |
NP_005938.1 |
3 - 3 |
326 |
121 |
GenBank mRNA |
AY168638.1 |
- |
3 - 3 |
327 |
121 |
View Full Product Details