Assay Design
Gene Transcripts
- Gene Symbol
- EMBP1
- Entrez Gene ID
-
647121
- Gene Name
- embigin pseudogene 1
- Gene Aliases
-
-
- Chromosome Location
-
Chr.1: 121519112 - 121571888 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_003955.1 |
|
8 - 8 |
2361 |
133 |
GenBank mRNA |
AK128714.1 |
- |
8 - 8 |
2361 |
133 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
SRGAP2 antisense RNA 1 |
SRGAP2-AS1 |
|
-170.9kb
|
Anti-sense |
Chr. - 121360156 - 121397905 on Build GRCh38 |
Downstream |
neuroblastoma breakpoint family member 17, pseudogene |
NBPF17P |
NBPF23, NBPF23P |
22.0mb
|
Anti-sense |
Chr. - 143595216 - 143635780 on Build GRCh38 |
View Full Product Details