Assay Design
Gene Transcripts
- Gene Symbol
- WDR26
- Entrez Gene ID
-
80232
- Gene Name
- WD repeat domain 26
- Gene Aliases
-
CDW2, GID7, MIP2
- Chromosome Location
-
Chr.1: 224385143 - 224434299 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AF130049.2 |
- |
3 - 3 |
2340 |
172 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AA731899.1 |
- |
2 |
|
|
Chr. 1 - 224173880 - 224429560 [+] on Build GRCh38 |
172 |
GenBankmRNA |
AI065023.1 |
- |
1 |
|
|
Chr. 1 - 224429297 - 224429978 [+] on Build GRCh38 |
172 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
microRNA 4742 |
MIR4742 |
mir-4742 |
-31.0kb
|
Sense |
Chr. - 224398227 - 224398311 on Build GRCh38 |
Downstream |
cornichon family AMPA receptor auxiliary protein 3 |
CNIH3 |
CNIH-3 |
3.2kb
|
Anti-sense |
Chr. - 224434640 - 224740549 on Build GRCh38 |
View Full Product Details