Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001004441.2 | NP_001004441.2 | 5 - 5 | 1968 | 133 |
XM_006714599.3 | XP_006714662.1 | 5 - 5 | 1730 | 133 | |
XM_011543366.2 | XP_011541668.1 | 5 - 5 | 1772 | 133 | |
XM_011543369.2 | XP_011541671.1 | 3 - 3 | 1728 | 133 | |
GenBank mRNA | BC137441.1 | - | 5 - 5 | 1747 | 133 |
BC137442.1 | - | 5 - 5 | 1744 | 133 | |
BC142957.1 | - | 4 - 4 | 1648 | 133 | |
CR749535.1 | - | 5 - 5 | 1762 | 133 |