Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001319075.1 | NP_001306004.1 | 2 - 2 | 1809 | 131 |
NM_006577.5 | NP_006568.2 | 2 - 2 | 1943 | 131 | |
GenBank mRNA | AK002009.1 | - | 1 - 1 | 1638 | 131 |
BC030579.2 | - | 2 - 2 | 1812 | 131 | |
BC047933.1 | - | 2 - 2 | 1942 | 131 |