Assay Design



Gene Transcripts

Gene Symbol
CEBPG
Entrez Gene ID
1054
Gene Name
CCAAT/enhancer binding protein gamma
Gene Aliases
GPE1BP, IG/EBP-1
Chromosome Location
Chr.19: 33373669 - 33382686 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001252296.1 NP_001239225.1 2 - 2 3463 134
NM_001806.3 NP_001797.1 2 - 2 3600 134
GenBank mRNA BC013128.1 - 2 - 2 3518 134

View Full Product Details