Assay Design
Gene Transcripts
- Gene Symbol
- DPPA3
- Entrez Gene ID
-
359787
- Gene Name
- developmental pluripotency associated 3
- Gene Aliases
-
STELLA
- Chromosome Location
-
Chr.12: 7711454 - 7717559 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_199286.3 |
NP_954980.1 |
4 - 4 |
996 |
131 |
GenBank mRNA |
AY317075.1 |
- |
4 - 4 |
996 |
131 |
View Full Product Details